Aluminium foil tapes, 425
: 3M
3M™ aluminum foil tape 425 is designed for numerous applications across a broad range of industries. This tape is comprised of a dead soft aluminium foil backing and coated with a highly engineered pressure sensitive acrylic adhesive.
- Thermally conductive tape helps dissipate heat and improves heating and cooling efficiency to protect temperature sensitive materials
- Resistant to chemicals to protect surfaces during de-paint processes and other chemical masking operations
- Improves heating and cooling efficiency to protect temperature sensitive materials
- Heat and light reflective tape protects surfaces and enhances lighting efficiency
- Withstands exposure to flame, weathering, moisture, and UV for a strong, long-lasting, and reliable bond
- Conforms to curved and uneven surfaces
- Tensile strength: 123 [Newton/25 mm]
- Aluminum foil back material
It can be used for a multitude of applications, such as masking, sealing, seaming, shielding, and chemical stripping.
This tape can also be used for harness wrapping, joining, electroplating, plastic protection, chemical milling, heat shielding, heat reflecting, and paint stripping.It reflects heat and light.
: Recognised by UL to UL 746C and classified by UL to UL 723, Class 'L' for low flammability rating.
Frequently Bought Together