Order Entry
Austria
ContactUsLinkComponent
Paraformaldehyde 32% (w/v) in aqueous solution methanol-free
Paraformaldehyde 32% (w/v) in aqueous solution methanol-free
Certificates
Paraformaldehyde 32% (w/v) in aqueous solution methanol-free
Certificates

Specification Test Results

  • Appearance (Color):
    Clear colorless
  • Form:
    Liquid
  • Ash:
    ≤0,01% (Paraformaldehyde content)
  • Comment:
    Manufactured from 95% Paraformaldehyde prills
  • Concentration:
    30,4 to 32% (Formaldehyde)
  • Trace Metal:
    Iron (Fe) ≤2,0% (Paraformaldehyde content)

Specification Test Results

Related Information

Frequently Bought Together