Order Entry
Austria
ContactUsLinkComponent
Multi-element ICH/USP parenteral combined standard Solution 2, Specpure®
Multi-element ICH/USP parenteral combined standard Solution 2, Specpure®
Catalog # 47369.AE
CAS Number:  
Multi-element ICH/USP parenteral combined standard Solution 2, Specpure®
Catalog # 47369.AE
CAS Number:  

Specifications

  • Matrix:
    15% HCl
  • Pk:
    100 mL
  • Description:
    Multi-Element ICH/USP Parenteral Combined Standard Solution 2

Specifications

About this item

7 components in 15% HCl matrix.

Concentration: Au 100µg/mL ±2%
Concentration: Ir 10µg/mL ±2%
Concentration: Os 10µg/mL ±2%
Concentration: Pd 10µg/mL ±2%
Concentration: Pt 10µg/mL ±2%
Concentration: Rh 10µg/mL ±2%
Concentration: Ru 10µg/mL ±2%