Order Entry
Austria
ContactUsLinkComponent
2,6-Di-tert-butyl-p-cresol ≥99%, GPR RECTAPUR®
2,6-Di-tert-butyl-p-cresol ≥99%, GPR RECTAPUR®
Supplier:  VWR Chemicals
Certificates
undefined
2,6-Di-tert-butyl-p-cresol ≥99%, GPR RECTAPUR®
Supplier:  VWR Chemicals
Certificates

Specification Test Results

  • Assay:
    Min. 99 %
  • Identification (IR):
    Conforms to structure
  • Freezing point:
    69.2 - 70.0 °C
  • Heavy metals (as Pb):
    Max. 10 ppm
  • Ignition residue:
    Max. 20 ppm
  • Ignition residue (SO4):
    Max. 0.1 %

Specification Test Results

Frequently Bought Together