Dual Lock™ SJ354D Reclosable Fastener Twin Pack
Supplier: 3M
Certificates
About this item
This reclosable fastener has a rubber based adhesive on the back of a black, polypropylene reclosable fastener. It's suitable for bonding many plastics, powder coated paints, and other low surface energy materials.
- Snap-in-place installation replaces screws, nails, snaps, and rivets
- Type 250 mates to Type 170, 250, or 400
- Closes with an audible snap
- Opens and closes 1000 times before losing 50% of original tensile strength
- Pressure-sensitive adhesive provides excellent initial tack to most surfaces
- Helps reduce vibration
Designed for use with many plastics (acrylic, polycarbonate, and AS), powder coated paints, and low surface energy materials (polypropylene and polyethylene). It's a type 250 stem density polyolefin fastener that is backed with a rubber based, pressure sensitive adhesive. The reclosable fastener has 40 mushroom shaped stems/cm² with a 5,8 mm engaged thickness. This product has low temperature performance, with a temperature resistance up to 50 °C long-term (70 °C short-term), and is therefore intended for indoor use only.
Frequently Bought Together