Order Entry
Austria
ContactUsLinkComponent
Pipette controller, manual
Pipette controller, manual
Catalog # HEATHS120479
Supplier:  HEATHROW
undefined
Pipette controller, manual
Catalog # HEATHS120479
Supplier:  HEATHROW

Specifications

  • Colour code:
    Blue
  • Capacity:
    1 - 100 ml
  • Description:
    Pipette controller, manual
  • Pk:
    1

Specifications

About this item

Comfortable to hold and easy to use, the manual inline pipette controller was designed with an ergonomic grip and bulb for simple and precise operation.

  • In-line ergonomic design reduces stress with ease of handling and operation
  • Conveniently accommodates either glass or plastic serological or volumetric pipettes ranging from 1 to 100 ml
  • Includes one replaceable 0,45 μm hydrophobic filter to protect your samples and internal components
  • Stay organised with two additional nose cones (green, white) for colour identification
  • Polypropylene nozzle and silicone adapter is autoclavable
  • Autoclavable (nose cone assembly)

A 'T' bar thumb lever controls the aspirating and dispensing speeds from serological pipettes from 1 to 100 ml in size.

Simply squeeze the bulb to create a vacuum to draw up fluid. Push the control lever backward (upward) to aspirate liquid through a pipette, or push the lever forward (downward) to dispense the liquid. A press of the central bulb on the unit blows-out any remaining fluid if necessary.

Dimension: 230×54×54 mm