Order Entry
Austria
ContactUsLinkComponent
 
 
 
Acetone, for GC Plus, Burdick & Jackson™
    
 
undefined
 
 
Acetone, for GC Plus, Burdick & Jackson™
    

 

  • Environmentally Preferable:
    Safer Human and Environmental Health
  • Environmental certification:
    Chem21 Solvent Selection Guide recommended after discussion

 

 

  • Assay (GC):
    Min. 99.8 %
  • Non-volatile matter:
    Max. 0.0005 %
  • Water (Karl Fischer):
    Max. 0.2 %
  • Suitability for residue analysis:
    Max. 5 ng/l
  • Suitability for GC-MS of dioxines, furanes, PCPs:
    complying

 

Frequently Bought Together