Order Entry
Austria
ContactUsLinkComponent
tert-Butyl methyl ether, Burdick & Jackson™
tert-Butyl methyl ether, Burdick & Jackson™
Catalog # HONC65810-25L
CAS Number:  
tert-Butyl methyl ether, Burdick & Jackson™
Catalog # HONC65810-25L
CAS Number:  

Specifications

  • Pack type:
    Metal drum
  • Pk:
    25 L
  • Seq:
    5327
  • Environmentally Preferable:

Specifications

Specification Test Results

  • Assay (GC):
    Min. 99 %
  • Non-volatile matter:
    Max. 0.002 %
  • Water (Karl Fischer):
    Max. 0.1 %
  • Free acid (as CH₃COOH):
    Max. 0.001 %
  • Aluminium (Al):
    Max. 0.00005 %
  • Boron (B):
    Max. 0.000002 %
  • Barium (Ba):
    Max. 0.00001 %
  • Calcium (Ca):
    Max. 0.00005 %
  • Cadmium (Cd):
    Max. 0.000005 %
  • Cobalt (Co):
    Max. 0.000002 %
  • Chromium (Cr):
    Max. 0.000002 %
  • Copper (Cu):
    Max. 0.000002 %
  • Iron (Fe):
    Max. 0.000005 %
  • Magnesium (Mg):
    Max. 0.00001 %
  • Manganese (Mn):
    Max. 0.000002 %
  • Nickel (Ni):
    Max. 0.000002 %
  • Lead (Pb):
    Max. 0.000005 %
  • Tin (Sn):
    Max. 0.00001 %
  • Zinc (Zn):
    Max. 0.000005 %
  • iso-Butyl methyl ketone:
    Max. 0.005 %
  • Carbonyl compound (as CO):
    Max. 0.001 %
  • Peroxides (as H₂O₂):
    Max. 0.00003 %
  • S-compounds (as S):
    Max. 0.00006 %

Specification Test Results

About this item

Synonyms: MTBE, 2-Methoxy-2-methylpropane, Methyl-t-Butyl Ether
 :
 :88.15 g/mol
Boiling Pt:55.3 °C
Density:0.74 g/cm3
Flash Pt:-28 °C
Flash Pt:–28 °C
UN:2398
ADR:3,II
 :GHS07
 :GHS02
CAS Number:1634-04-4
Formula:(CH₃)₃COCH₃
MDL Number:MFCD00008812