Order Entry
Austria
ContactUsLinkComponent
Acetonitrile Reag. Ph. Eur., gradient grade for HPLC, Burdick & Jackson™
Flammable
Irritant
Acetonitrile Reag. Ph. Eur., gradient grade for HPLC, Burdick & Jackson™
   Sustainable Options Available
Certificates
undefined
Flammable
Irritant
Acetonitrile Reag. Ph. Eur., gradient grade for HPLC, Burdick & Jackson™
   Sustainable Options Available

Specifications

  • Environmentally Preferable:
    Safer Human and Environmental Health
  • Environmental certification:
    EPA Safer Chemical Solvent (Green Circle Rating); Chem21 Solvent Selection Guide recommended by default and after discussion

Specifications

Specification Test Results

  • Appearance:
    complying
  • Assay (GC):
    Min. 99.9 %
  • Boiling range 80-82 °C:
    Min. 95 %
  • Density (D 20/20):
    0.782 - 0.784
  • Refractive index (n 20/D):
    1.3435 - 1.3445
  • Non-volatile matter:
    Max. 0.0002 %
  • Water (Karl Fischer):
    Max. 0.01 %
  • Acidity:
    Max. 0.0002 meq/g
  • Alkalinity:
    Max. 0.0001 meq/g
  • Transmittance at 195 nm:
    Min. 75 %
  • Transmittance at 200 nm:
    Min. 93 %
  • Transmittance from 230 nm:
    Min. 99 %
  • HPLC Gradient at 210 nm:
    Max. 3 mAU
  • HPLC Gradient at 254 nm:
    Max. 0.5 mAU
  • Baseline drift at 210 nm:
    Max. 15 mAU
  • Fluorescence (chinin) at 254 nm:
    Max. 1 ppb
  • Fluorescence (chinin) at 365 nm:
    Max. 0.5 ppb
  • Phthalates as dibutylphthalate:
    Max. 0.1 mg/l
  • Reaction of the 10 % solution to Litmus:
    complying

Specification Test Results

Frequently Bought Together