Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Austria
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
HistoPerls Kit
HistoPerls Kit Highlighting hemosiderin, pathological pigment (ferric salts)
HistoPerls Kit Highlighting hemosiderin, pathological pigment (ferric salts)
HistoPerls Kit Highlighting hemosiderin, pathological pigment (ferric salts)
Catalog # 720-0359
Supplier:
RAL
CAS Number:
Expand image
HistoPerls Kit Highlighting hemosiderin, pathological pigment (ferric salts)
Catalog # 720-0359
Supplier:
RAL
CAS Number:
Specifications
Pk:
1 KIT
Seq:
0075
Specifications
About this item
Product Details
:
:
1
Product Details