Order Entry
Austria
ContactUsLinkComponent
 
Silicone oil, GPR RECTAPUR® for oil baths up to 250°C
 
undefined
Silicone oil, GPR RECTAPUR® for oil baths up to 250°C
 

 

  • Density (20/4):
    0.966 - 0.972
  • Viscosity (25°C):
    330 - 370 cSt

 

Frequently Bought Together