Order Entry
Austria
ContactUsLinkComponent
 
Corrosive
Cerium(IV) sulphate 0,05 M volumetric solution, Fluka™
Catalog # 34253-1L
CAS Number:  
Corrosive
Cerium(IV) sulphate 0,05 M volumetric solution, Fluka™
Catalog # 34253-1L
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    1 L
  • Seq:
    5037

Specifications

Specification Test Results

  • Factor:
    0.998 - 1.002

Specification Test Results

About this item

 :
 :332.24 g/mol
Storage Temperature:Ambient
UN:3264
ADR:8,III
 :GHS05
CAS Number:13590-82-4
Formula:Ce(SO₄)₂
MDL Number:MFCD00148852