VWR®, Lockable contact plate for pharmaceutical industries
Supplier: VWR Chemicals
New Product
Certificates
About this item
These contact plates, with triple-bagged sterile packaging and lockable lid, are ideal for environmental control of air, surfaces, gloves and overalls inside restricted areas like isolators and cleanrooms. The composition of the base culture medium complies with the recommendations of the current United States, European and Japanese Pharmacopoeia.
- The locking lid allows an easy locking of plates after sampling, adding security during transportation
- 9 month shelf life and room temperature storage ensure easy storage and the maintaining of culture efficiency for a long time
- A data matrix barcode allows identification of every plate used for environmental control
- The complete triple-bagged packaging is sterile to ensure that when entering a clean area it avoids microbiological contamination
- Neutralising agents included in the medium inactivate residual disinfectants that could reduce the real count of live microorganisms
- Different lid types available: Screw on or press on (see the videos for more details)
- For plates with screw-on lid: lid fixed on CLOSE position allows safe transport avoiding risk of contamination due to its possible opening during the transport; lid fixed on AIR position, allows AEROBIC and ANAEROBIC incubations
Related Information
Certificates
Search by Lot NumberLockable Contact Plate with screw-on lid for Pharmaceutical Industries, VWR® Lockable Contact Plates with press-on lid for Pharmaceutical Industries Lockable Contact Plates with screw-on lid for Pharmaceutical Industries
Videos
SDS_EN_PL_VWRC15203VS.pdf SDS_EN_UK_VWRC15203VS.pdf SDS_EN_UK_VWRCC30884.pdf SDS_EN_UK_VWRCC31114PY.pdf SDS_EN_UK_VWRCC31114RI.pdf
SDS
More
spec_15203VS spec_15204VS spec_C30884CY spec_C30884TI spec_C31114PY
Product specification
More
Frequently Bought Together