1,4-Dioxane, anhydrous 99.80%, AcroSeal®
Supplier: Thermo Fisher Scientific
Certificates
About this item
Synonyms: Glycolethylether, 1,4-Dioxacyclohexane, Dioxane, p-Dioxane, 1,4-Diethylene dioxide
CAS Number:123-91-1
:
Formula:C₄H₈O₂
MDL Number:MFCD00006571
:88.11 g/mol
Boiling Pt:101 °C
Melting Pt:12 °C
Density:1.034
Flash Pt:12°C
Storage Temperature:Ambient
UN:1165
ADR:3,II
:Packaged under argon.
EINECS:204-661-8
:GHS02
:GHS07
:GHS08
Specifications
- Environmentally Preferable:
Specifications
Specification Test Results
- Appearance:Colorless liquid
- Assay (GC):≥99.75%
- Water content (Karl Fischer Titration):≤50 ppm
- Residue on evaporation:≤5 ppm
Specification Test Results
Frequently Bought Together