Gas-safety adapter 360 for Flame 100
WART2.000.701
: WARTEWIG
- Description:Gas-safety adapter 360 for Flame 100
- Environmentally Preferable:
- Pk:1
Gas-safety adapter 360 for Flame 100.
- With integrated pressure regulator
- Continuous cartridge operation 65 minutes
Ideal for cartridges CV 360, 52 g from Camping gas.