Chemical resistant gloves, Showa KV660
Supplier: Showa
Certificates
About this item
Providing defense against a broad range of hazards these cut and chemical-resistant gloves are forged with triple-dipped PVC and a reinforced Kevlar® liner.
- Rough surface finish offers a good grip and increased durability
- Even after multiple washes, the KV660 maintains its barrier to hazards
- Seamless Aramid/Kevlar® knit
The KV660 provides first-class defense against oils, chemicals, abrasion, and cuts.
Typical applications include: Chemical spray and treatment, coating preparation, painting & spray workshops, bottling, HVAC, plumbing, guttering and drain pipe, drainage, piping.
Certifications: EN 388:2016 (4X42B); EN 374-1:2016 Type B (JKL); EN 374-5:2016; Cat. III
Packaging: 10 pairs per polybag.
Specifications
- Catalog No:
- 112-5998
- 112-5996
- 112-5995
- 112-5997
- Model:
- Showa KV660
- Showa KV660
- Showa KV660
- Showa KV660
- Ambidextrous / hand-specific:
- Hand-specific
- Hand-specific
- Hand-specific
- Hand-specific
- Disposable / reusable:
- Reusable
- Reusable
- Reusable
- Reusable
- Liner material:
- Kevlar®
- Kevlar®
- Kevlar®
- Kevlar®
- Finish:
- Rough surface
- Rough surface
- Rough surface
- Rough surface
- Cuff style:
- Pinked
- Pinked
- Pinked
- Pinked
- EN norm:
- EN 388:2016 (4X42B); EN 374-1:2016 Type B (JKL); EN 374-5:2016
- EN 388:2016 (4X42B); EN 374-1:2016 Type B (JKL); EN 374-5:2016
- EN 388:2016 (4X42B); EN 374-1:2016 Type B (JKL); EN 374-5:2016
- EN 388:2016 (4X42B); EN 374-1:2016 Type B (JKL); EN 374-5:2016