ContactUsLinkComponent
General purpose gloves, Showa 7166R
General purpose gloves, Showa 7166R
Catalog # 112-5892
Supplier:  Showa
undefined
General purpose gloves, Showa 7166R
Catalog # 112-5892
Supplier:  Showa
Specifications
  • Glove size:
    10
  • Ambidextrous / hand-specific:
    Hand-specific
  • Disposable / reusable:
    Reusable
  • Colour:
    Dark blue
  • Length:
    260 mm
  • Model:
    7166R
  • EN norm:
    EN 388:2016 (4111X)
  • Cuff style:
    Safety cuff
  • Liner material:
    Cotton jersey
  • Finish:
    Full nitrile coating
  • Pk:
    1 PAIR
Specifications
About this item
The 7166 general purpose gloves feature an impermeable full nitrile coating that protects hands from a range of hazards, including abrasion, oils, and chemicals.
  • Reinforced safety cuff prevents liquid and debris from entering the glove
  • Soft cotton liner controls the temperature and wicks away sweat
  • Rough surface grip is ideal for wet or greasy environments and increased protection
Typical applications include: Chemical spray and treatment, cutting roses and thorns, public utility, washing and cleaning, drainage, piping, guttering and drain pipe, plumbing, water/sewage treatment.
Certifications: EN 388:2016 (4111X); Cat. II