ContactUsLinkComponent
General purpose gloves, Showa 376R
General purpose gloves, Showa 376R
Supplier:  Showa
Certificates
General purpose gloves, Showa 376R
Supplier:  Showa
Specifications
  • Catalog No:
  • 112-5805
  • 112-5806
  • 112-5803
  • 112-5807
  • 112-5804
  • Model:
  • Showa 376R
  • Showa 376R
  • Showa 376R
  • Showa 376R
  • Showa 376R
  • Ambidextrous / hand-specific:
  • Hand-specific
  • Hand-specific
  • Hand-specific
  • Hand-specific
  • Hand-specific
  • Disposable / reusable:
  • Reusable
  • Reusable
  • Reusable
  • Reusable
  • Reusable
  • Liner material:
  • Nylon/polyester
  • Nylon/polyester
  • Nylon/polyester
  • Nylon/polyester
  • Nylon/polyester
  • Finish:
  • 3/4 nitrile dipped with extra foam nitrile on palm
  • 3/4 nitrile dipped with extra foam nitrile on palm
  • 3/4 nitrile dipped with extra foam nitrile on palm
  • 3/4 nitrile dipped with extra foam nitrile on palm
  • 3/4 nitrile dipped with extra foam nitrile on palm
  • Cuff style:
  • Knitted elasticated wrist and cuff
  • Knitted elasticated wrist and cuff
  • Knitted elasticated wrist and cuff
  • Knitted elasticated wrist and cuff
  • Knitted elasticated wrist and cuff
  • EN norm:
  • EN 388:2016 (4121X)
  • EN 388:2016 (4121X)
  • EN 388:2016 (4121X)
  • EN 388:2016 (4121X)
  • EN 388:2016 (4121X)