ContactUsLinkComponent
Centrifugal devices for RNA and DNA extraction, NAB Nanosep®
Centrifugal devices for RNA and DNA extraction, NAB Nanosep®
Certificates
undefined
Centrifugal devices for RNA and DNA extraction, NAB Nanosep®
Specifications
  • Catalog No:
  • 516-9341
  • PALLODNABC33
  • PALLODNABC34
  • Filtration area:
  • 0.28 cm²
  • 0.28 cm²
  • Max. RCF:
  • 14,000 ×g
  • 14,000 ×g
  • Ø×H:
  • 45×23 mm
  • 45×23 mm
  • Sample volume:
  • max. 500 µl
  • max. 500 µl
  • Temperature:
  • 15 - 30 °C (59 - 86 °F)
  • 15 - 30 °C (59 - 86 °F)