Dosing spoon and measuring spoon set
About this item
Dosing and measuring spoons can be used to measure small amounts of powder, granulates and liquids exactly and dose them precisely. For precise measuring, excess medium is stripped off the upper edge of the spoon. The content volume is engraved on the handle.
- Content volume: 0,5; 1,0; 2,5; 5,0; 10; 15; 25; and 50 ml
- Set of volumetric spoons with eight sizes
- PS
- With stripper edge
The flat front edge of the volumetric spoon is ideal for picking up medium from the bottom of the container or for getting into the corners of the container, thus allowing for good emptying of small remaining amounts. The flat base makes it possible to set the volumetric spoon down without spilling the content.
An eyelet in the end of the handle can be used to hang up a spoon or to bundle several together.
Certifications: Complies with EU food requirements and FDA regulations.
2 Options Available Below
Product Details & Documents
Dosing and measuring spoons can be used to measure small amounts of powder, granulates and liquids exactly and dose them precisely. For precise measuring, excess medium is stripped off the upper edge of the spoon. The content volume is engraved on the handle.
- Content volume: 0,5; 1,0; 2,5; 5,0; 10; 15; 25; and 50 ml
- Set of volumetric spoons with eight sizes
- PS
- With stripper edge
The flat front edge of the volumetric spoon is ideal for picking up medium from the bottom of the container or for getting into the corners of the container, thus allowing for good emptying of small remaining amounts. The flat base makes it possible to set the volumetric spoon down without spilling the content.
An eyelet in the end of the handle can be used to hang up a spoon or to bundle several together.
Certifications: Complies with EU food requirements and FDA regulations.
Product Family Options
Product Information