Orders LinkContactUsLinkComponent
Detectable forceps, sterile
Detectable forceps, sterile
Supplier:   BURKLE
Detectable forceps, sterile
Supplier:  BURKLE
Certificates
Detectable forceps, sterile
Detectable forceps, sterile
3 Options Available Below
Product Details & Documents
These samplers are not only coloured blue but also have a special additive in the material that makes them visible to metal detectors or X rays. They can, therefore be rejected with standard systems for checking for foreign objects, even though they are made of plastic. These disposable forceps are suitable for precise lab work and are ideal for grasping, lifting and moving small objects, for example, fine tissue, small items or solid samples.
  • PS detectable, blue
  • Individually packed and sterilised
  • For single use
  • Cleanroom manufactured (Class 7)
  • Overall length 130 mm
The fluted handles ensure optimum and secure handling as well as precise work. These light plastic single-use forceps for allow for fatigue-free work. The cleanroom manufactured lab assistants meet the highest standards preventing cross-contamination or impurities in the sample being examined.
These lab forceps are suitable for chemical, pharmaceutical or cosmetic applications as well as the food industry and scientific uses.
Pointed/angled shape: These disposable forceps have fine, smooth, curved gripping tips. Different materials can be held and processed reliably with the pointed ends.
Pointed shape: These anatomical forceps have a straight tip, fluted on the inside making it possible to grip sensitive materials reliably.
Broad tip shape: These disposable forceps have ends with a broad tip. The broad tip is smooth and blunt preventing the material from being damaged when gripped.
Certifications: Complies with EU food requirements and FDA regulations.
Product Family Options
Product Information
  • Type
    Colour
    Sterile
    Length
    Packed
    Availability
    Price
  • Pointed, angled
    Blue
    +
    130 mm
    Individually
    Log in for availability
    Log in for availability
    1 * 100 ST
    1 * 100 ST - 92,20 €
    92,20 €
    1 * 100 ST
       92,20 €
  • Pointed, straight
    Blue
    +
    130 mm
    Individually
    Log in for availability
    Log in for availability
    1 * 100 ST
    1 * 100 ST - 92,20 €
    92,20 €
    1 * 100 ST
       92,20 €
  • Broad tip, straight
    Blue
    +
    130 mm
    Individually
    Log in for availability
    Log in for availability
    1 * 100 ST
    1 * 100 ST - 92,20 €
    92,20 €
    1 * 100 ST
       92,20 €