Desiccator lids, DURAN®
Supplier: DWK Life Sciences
Certificates
About this item
DURAN®, clear borosilicate glass, 3.3. Suitable for WERTEX type vacuum desiccators.
- With special tube (NS 24/29, Type WERTEX)
- With ring-grooved flange
Frequently Bought Together