Order Entry
Austria
ContactUsLinkComponent
Propylene carbonate ≥99.0% (by GC), GPR RECTAPUR®
Propylene carbonate ≥99.0% (by GC), GPR RECTAPUR®
Supplier:  VWR Chemicals
Certificates
undefined
Propylene carbonate ≥99.0% (by GC), GPR RECTAPUR®
Supplier:  VWR Chemicals
Certificates
Limited Time Sale
Select items below are currently on sale

Specifications

  • Environmentally Preferable:

Specifications

Specification Test Results

  • Assay (GC):
    ≥ 99,0%
  • Identity (IR):
    passes test
  • Density (d 20 °C/4 °C):
    1,204 - 1,208
  • Appearance:
    clear liquid

Specification Test Results

Frequently Bought Together