Orders LinkContactUsLinkComponent
Forceps, disposable | Avantor
Forceps, disposable
Anbieter:   BURKLE
Forceps, disposable
Anbieter:  BURKLE
Zertifikate
Forceps, disposable
6 Optionen unten verfügbar
Product Details & Documents
The disposable forceps are suitable for precise lab work and are ideal for grasping, lifting and moving small objects, for example fine tissue, small items or solid samples.
  • PS, white
  • Can be individually packed and sterilised
  • For single use
  • Clean room manufactured (Class 7 cleanroom)
  • Overall length 130 mm
The fluted handles ensure optimum and secure handling as well as precise work. Light plastic forceps for single use allow for fatigue-free work. Cleanroom manufactured lab assistants meet the highest standards and prevent cross-contamination or impurities in the sample being examined.
These lab forceps are suitable for chemical, pharmaceutical or cosmetic applications as well as the food industry and scientific uses.
Pointed/angled shape: These disposable forceps have fine, smooth, curved gripping tips. Different materials can be held and processed reliably with the pointed ends.
Pointed shape: These anatomical forceps have a straight tip, fluted on the inside making it possible to grip sensitive materials reliably.
Broad tip shape: These disposable forceps have ends with a broad tip. The broad tip is smooth and blunt preventing material from being damaged when gripped.
Zertifizierung: Complies with EU food requirements and FDA regulations.
Verpackung: Available either in a large pack with 100 pieces or 100 pieces packed individually and radiation sterilised.
Product Family Options
Produktinformationen
  • Type
    Sterile
    Packed
    Verfügbarkeit
    Preis
  • Pointed, angled
    -
    Large pack
    Einloggen für Verfügbarkeit
    Einloggen für Verfügbarkeit
    1 * 100 ST
    1 * 100 ST - 37,30 €
    37,30 €
    1 * 100 ST
       37,30 €
  • Pointed, angled
    +
    Individually
    Einloggen für Verfügbarkeit
    Einloggen für Verfügbarkeit
    1 * 100 ST
    1 * 100 ST - 62,00 €
    62,00 €
    1 * 100 ST
       62,00 €
  • Pointed, straight
    -
    Large pack
    Einloggen für Verfügbarkeit
    Einloggen für Verfügbarkeit
    1 * 100 ST
    1 * 100 ST - 37,30 €
    37,30 €
    1 * 100 ST
       37,30 €
  • Pointed, straight
    +
    Individually
    Einloggen für Verfügbarkeit
    Einloggen für Verfügbarkeit
    1 * 100 ST
    1 * 100 ST - 62,00 €
    62,00 €
    1 * 100 ST
       62,00 €
  • Broad tip, straight
    -
    Large pack
    Einloggen für Verfügbarkeit
    Einloggen für Verfügbarkeit
    1 * 100 ST
    1 * 100 ST - 37,30 €
    37,30 €
    1 * 100 ST
       37,30 €
  • Broad tip, straight
    +
    Individually
    Einloggen für Verfügbarkeit
    Einloggen für Verfügbarkeit
    1 * 100 ST
    1 * 100 ST - 62,00 €
    62,00 €
    1 * 100 ST
       62,00 €