Chemical resistant coveralls, AllSafe, Type 5+6, SAFE®
Supplier: DACH SCHUTZBEKLEIDUNG
Certificates
About this item
The AllSafe coverall combines protection, wearing comfort, and excellent price-performance ratio.
- Breathable nonwoven fabric
- Antistatic material
- Self-adhesive zipper cover
- Broad size range
The unique material is antistatic, lint-free, liquid- and spray-tight. Thus, the coverall is suited for various fields of application, ranging from industries such as chemistry and engineering to the pharmaceutical and medical sector, but applications may also include craft, agriculture, and landscaping.
Certifications: Meets the requirements for a chemical protective coverall Type 5 and 6. Also complies with the European standards EN 1073-2 and 1149-5.
Specifications
- Catalog No:
- DACH571/XL
- DACH571/M
- DACH571/XXXL
- DACH571/XXL
- DACH571/L
- Anti-static:
- +
- +
- +
- +
- +
- Garment application:
- Chemical protection, General Purpose, Medical
- Chemical protection, General Purpose, Medical
- Chemical protection, General Purpose, Medical
- Chemical protection, General Purpose, Medical
- Chemical protection, General Purpose, Medical