Order Entry
Austria
ContactUsLinkComponent
N,O-Bis(trimethylsilyl)trifluoroacetamide with 1% trimethylchlorosilane, Cerilliant® for GC derivatisation, analytical standard, Supelco®
Flammable
Corrosive
N,O-Bis(trimethylsilyl)trifluoroacetamide with 1% trimethylchlorosilane, Cerilliant® for GC derivatisation, analytical standard, Supelco®
Catalog # B-023-10X1ML
Supplier:  Merck
CAS Number:  
Flammable
Corrosive
N,O-Bis(trimethylsilyl)trifluoroacetamide with 1% trimethylchlorosilane, Cerilliant® for GC derivatisation, analytical standard, Supelco®
Catalog # B-023-10X1ML
Supplier:  Merck
CAS Number:  

Specifications

  • Pk:
    10
  • Seq:
    2076
  • Pk Info:
    10× 1 ml

Specifications

About this item

 :
 :257.40 g/mol
Storage Temperature:Freezer
UN:2920
ADR:8(3),II
 :GHS02
 :GHS05
CAS Number:25561-30-2
Formula:C₈H₁₈F₃NOSi₂
MDL Number:MFCD00008269