Order Entry
Austria
ContactUsLinkComponent
 
 
 
Acetonitrile ≥99.8% (by GC) for preparative HPLC, Riedel-de Haën™
    
 
undefined
 
 
Acetonitrile ≥99.8% (by GC) for preparative HPLC, Riedel-de Haën™
    

 

  • Environmentally Preferable:
    Safer Human and Environmental Health
  • Environmental certification:
    Chem21 Solvent Selection Guide recommended by default and after discussion

 

 

  • Assay (GC):
    Min. 99.8 %
  • Non-volatile matter:
    Max. 0.0001 %
  • Water (Karl Fischer):
    Max. 0.1 %
  • Free alkali (as NH₃):
    Max. 0.0005 %
  • Transmittance at 220 nm:
    Min. 50 %
  • Transmittance from 240 nm:
    Min. 99 %
  • Explosion Limit:
    16 %
  • Autoignition Temperature:
    973 °F
  • Vapor Pressure:
    72.8 mmHg ( 20 °C)

 

Frequently Bought Together