Order Entry
Austria
ContactUsLinkComponent
Bis(2-methoxyethyl)ether ≥99.5% (by GC), Puriss. p.a., Riedel-de Haën™
Bis(2-methoxyethyl)ether ≥99.5% (by GC), Puriss. p.a., Riedel-de Haën™
Catalog # HONC32210-190KG
CAS Number:  
Bis(2-methoxyethyl)ether ≥99.5% (by GC), Puriss. p.a., Riedel-de Haën™
Catalog # HONC32210-190KG
CAS Number:  

Specifications

  • Pack type:
    Steel drum
  • Pk:
    190 kg
  • Seq:
    4672

Specifications

Specification Test Results

  • Identity (NMR):
    complying
  • Assay (GC):
    min. 99.5 %
  • Refractive index (n 20/D):
    1.407 - 1.409
  • Water (Karl Fischer):
    max. 0.1 %
  • Peroxides (as H2O2):
    max. 50 ppm
  • Aluminium (Al):
    max. 0.5 ppm
  • Barium (Ba):
    max. 0.1 ppm
  • Bismuth (Bi):
    max. 0.1 ppm
  • Calcium (Ca):
    max. 0.5 ppm
  • Cadmium (Cd):
    max. 0.05 ppm
  • Cobalt (Co):
    max. 0.02 ppm
  • Chromium (Cr):
    max. 0.02 ppm
  • Copper (Cu):
    max. 0.02 ppm
  • Iron (Fe):
    max. 0.1 ppm
  • Potassium (K):
    max. 0.5 ppm
  • Lithium (Li):
    max. 0.1 ppm
  • Magnesium (Mg):
    max. 0.1 ppm
  • Manganese (Mn):
    max. 0.02 ppm
  • Molybdenum (Mo):
    max. 0.1 ppm
  • Sodium (Na):
    max. 0.5 ppm
  • Nickel (Ni):
    max. 0.02 ppm
  • Lead (Pb):
    max. 0.1 ppm
  • Strontium (Sr):
    max. 0.1 ppm
  • Zinc (Zn):
    max. 0.1 ppm

Specification Test Results

About this item

 :
Boiling Pt:162 °C
Melting Pt:–64 °C
Density:0.9451 g/cm3 (20 °C)
Flash Pt:57 °C
Storage Temperature:Ambient
EINECS:203-924-4
UN:3271
ADR:3,III
 :134.18 g/mol
 :GHS08
 :GHS02
CAS Number:111-96-6
Formula:C₆H₁₄O₃
MDL Number:MFCD00008503