Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Austria
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Buffers
pH Calibration Buffers
Buffer solution, pH 2 (20 °C), Fluka™
Buffer solution, pH 2 (20 °C), Fluka™
Buffer solution, pH 2 (20 °C), Fluka™
456098-500ML
:
Honeywell Chemicals
:
Buffer solution, pH 2 (20 °C), Fluka™
456098-500ML
:
Honeywell Chemicals
:
Pack type:
Plastic bottle
Pk:
500 mL
Colour:
Neutral
pH accuracy:
±0.02
pH value:
2,00