

Potassium hydroxide 0.1 M (0.1 N) in ethanol volumetric solution, Fluka™
Supplier: Honeywell Chemicals
Certificates
About this item
CAS Number:1310-58-3
:
Formula:KOH
MDL Number:MFCD00003553
:56.11 g/mol
Flash Pt:14 °C
UN:1170
ADR:3,II
:GHS07
:GHS02
Storage Temperature:Ambient
Specification Test Results
- Factor:0.999 - 1.001
- BAM/EMPA reference material:effective
- BAM/EMPA Certificate Number:effective
- Traceable to NIST SRM:effective
Specification Test Results
Frequently Bought Together