Human recombinant KRAS,His Tag N-Terminus
Catalog # ABCAAB96817-100
Supplier: Abcam
CAS Number:
Specifications
- Pk:100 µG
- Protein/peptide type:Recombinant
- Species:Human
- Tag sequence:His tag N-Terminus
- Storage conditions:Store at –20 or –80 °C
- Accession no.:P01116-2
- Protein/peptide name:KRAS
- Sequence:MGSSHHHHHH SSGLVPRGSH MTEYKLVVVG AGGVGKSALT IQLIQNHFVD EYDPTIEDSY RKQVVIDGET CLLDILDTAG HEEYSAMRDQ YMRTGEGFLC VFAINNTKSF EDIHHYREQI KRVKDSEDVP MVLVGNKCDL PSRTVDTKQA QDLARSYGIP FIETSAKTRQ GVDDAFYTLV REIRKHKEKM SKDGKKKKKK SKTKC
- Concentration:100 µg at 0,5 mg/ml
500 µg at 0,5 mg/ml
Specifications
About this item
Caution:
All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.