Order Entry
Austria
ContactUsLinkComponent
(S)-2-((5-Fluoro-2,4-dinitrophenyl)amino)propanamide ≥99.0%, LiChropur™ for chiral derivatisation, Supelco®
Irritant
(S)-2-((5-Fluoro-2,4-dinitrophenyl)amino)propanamide ≥99.0%, LiChropur™ for chiral derivatisation, Supelco®
Catalog # 71478-50MG
Supplier:  Merck
CAS Number:  
undefined
Irritant
(S)-2-((5-Fluoro-2,4-dinitrophenyl)amino)propanamide ≥99.0%, LiChropur™ for chiral derivatisation, Supelco®
Catalog # 71478-50MG
Supplier:  Merck
CAS Number:  

Specifications

  • Pk:
    50 mg
  • Seq:
    2370

Specifications

About this item

 :
 :272.192 g/mol
 :M-Clarity™ quality level = MQ100
 :GHS07
Storage Temperature:Refrigerator
CAS Number:95713-52-3
Formula:C₉H₉FN₄O₅
MDL Number:MFCD00042049