
(-)-ɑ-Methoxy-ɑ-(trifluoromethyl)phenylacetyl chloride ≥99.0%, LiChropur™ for chiral derivatisation, Supelco®
Supplier: Merck
Certificates
About this item
CAS Number:39637-99-5
:
Formula:C₁₀H₈ClF₃O₂
MDL Number:MFCD00044400
:252.62 g/mol
ADR:8,II
:M-Clarity™ quality level = MQ100
UN:3265
:GHS05
Frequently Bought Together