
Valinomycin, Selectophore™, function tested ionophore, Supelco®
Catalog # 60403-100MG
Supplier: Merck
CAS Number:
Specifications
- Pk:100 mg
- Seq:2440
Specifications
About this item
:
:1111.34 g/mol
ADR:6.1,I
:M-Clarity™ quality level = MQ100
UN:2811
:GHS06
CAS Number:2001-95-8
Formula:C₅₄H₉₀O₁₈N₆
MDL Number:MFCD00005114