N-Isobutyryl-L-cysteine ≥97.0%, LiChropur™ for chiral derivatisation, Supelco®
Supplier: Merck
Certificates
About this item
CAS Number:124529-02-8
:
Formula:C₇H₁₃NO₃S
MDL Number:MFCD00155635
:191.25 g/mol
:M-Clarity™ quality level = MQ100
Frequently Bought Together