Orders LinkContactUsLinkComponent
pH indicator papers, Supelco®
pH indicator papers, Supelco®
pH indicator papers, Supelco®
Supplier:  Merck
Certificates
pH indicator papers, Supelco®
3 Options Available Below
Product Details & Documents
Traditional pH indicator test papers, supplied on a 4,8 metre roll, consisting of high quality filter paper impregnated with indicator and mixed indicator solutions then dried and cut to size.
  • Roll format are easy to use and most common ranges are available in a refill format
  • Quality assurance - tests are checked and calibrated against certified buffer solutions
  • Wide and narrow pH ranges available
Specifications for Product Family
Specifications
  • Catalog No:
  • 1.09560.0003
  • 1.09562.0003
  • 1.09564.0003
  • M-Clarity quality level:
  • MQ100
  • MQ100
  • MQ100
Specifications
Product Family Options
Product Information
  • pH range
    pH gradation
    Availability
    Price
  • Acilit®
    0,5 - 5,0
    0,5
    Log in for availability
    Log in for availability
    1 * 3 Rolle
    1 * 3 Rolle - 50,50 €
    50,50 €
    1 * 3 Rolle
       50,50 €
    Specifications:
    More
    Specifications
    Cat. No.
    1.09560.0003
    M-Clarity quality level
    MQ100
  • Alkalit®
    9,5 - 13,0
    0,5
    Log in for availability
    Log in for availability
    1 * 3 Rolle
    1 * 3 Rolle - 55,40 €
    55,40 €
    1 * 3 Rolle
       55,40 €
    Specifications:
    More
    Specifications
    Cat. No.
    1.09562.0003
    M-Clarity quality level
    MQ100
  • Neutralit®
    5,5 - 9,0
    0,5
    Log in for availability
    Log in for availability
    1 * 3 Rolle
    1 * 3 Rolle - 51,30 €
    51,30 €
    1 * 3 Rolle
       51,30 €
    Specifications:
    More
    Specifications
    Cat. No.
    1.09564.0003
    M-Clarity quality level
    MQ100