Orders LinkContactUsLinkComponent
Ionic strength adjustor buffer solution for fluoride electrode, Supelco®
Ionic strength adjustor buffer solution for fluoride electrode, Supelco®
Ionic strength adjustor buffer solution for fluoride electrode, 1,4% HOAc, 8,2% NaOAc, 5,8% NaCl, trace CDTA), Plastic bottle, 475 mL
Catalog # 443123-475ML
Supplier:   Merck
Ionic strength adjustor buffer solution for fluoride electrode, Supelco®
Catalog # 443123-475ML
Supplier:  Merck
CAS Number:  
Ionic strength adjustor buffer solution for fluoride electrode, Supelco®
Ionic strength adjustor buffer solution for fluoride electrode, 1,4% HOAc, 8,2% NaOAc, 5,8% NaCl, trace CDTA), Plastic bottle, 475 mL
CAS Number:  
Price
Inquire For Price
Log in for availability
1 * 475 mL
Product Details & Documents
It may be used as TISAB buffer in determination of fluoride in coal, coal fly ash, phosphate rock, limestone, mineral clay, fossilised materials, oyster tissue and vegetation using pyrohydrolysis for sample decomposition.
Specifications
Specifications
  • Pack type:
    Plastic bottle
  • Pk:
    475 mL
  • Description:
    Ionic strength adjustor buffer solution for fluoride electrode
  • Contents:
    1,4% HOAc, 8,2% NaOAc, 5,8% NaCl, trace CDTA)
Specifications