TISAB IV Solution for Determination of Fluoride, Supelco®
Lieferant: Merck
Zertifikate
Über diesen Artikel
Total ionic strength adjustment buffers (TISAB), is generally used for the microdetermination of fluorides.
TISAB IV was used as a metal ion complexing and ionic concentration adjustment agent, in an experiment investigating the increase in fluoride release, from glass ionomer cement, after coating the discs of glass ionomer with 10% silver fluoride (AgF).
Spezifikationen
- M-Clarity quality level:MQ100
Spezifikationen
Frequently Bought Together