Order Entry
Austria
ContactUsLinkComponent
Phosphate buffer solution, BioUltra, Supelco®
Phosphate buffer solution, BioUltra, Supelco®
Catalog # 76847-100ML-F
Supplier:  Merck
CAS Number:  
Phosphate buffer solution, BioUltra, Supelco®
Catalog # 76847-100ML-F
Supplier:  Merck
CAS Number:  

Specifications

  • Concentration:
    >0,044 M Na₂HPO₄, >0,022 M NaH₂PO₄
  • Pk:
    100 mL
  • Description:
    Phosphate buffer
  • M-Clarity quality level:
    MQ100
  • pH accuracy:
    ±0.2
  • pH value:
    7,2

Specifications

About this item