Order Entry
Austria
ContactUsLinkComponent
Mag-Bind® FFPE DNA/RNA 96 Kit
Mag-Bind® FFPE DNA/RNA 96 Kit
Supplier:  OMEGA BIO-TEK
Certificates
undefined
Mag-Bind® FFPE DNA/RNA 96 Kit
Supplier:  OMEGA BIO-TEK

Specifications

  • Method / Format:
    Magnetic beads
  • Prep size:
    Mini
  • Refine target molecule:
    Total RNA
    Total DNA
  • Sample type:
    FFPE tissues
  • Target molecule:
    Copurification of DNA & RNA
    DNA
    RNA

Specifications

Related Information

Frequently Bought Together