Order Entry
Austria
ContactUsLinkComponent
ɑ-Cyano-4-hydroxycinnamic acid, Ultrapure matrix substance for MALDI-MS, Supelco®
Irritant
ɑ-Cyano-4-hydroxycinnamic acid, Ultrapure matrix substance for MALDI-MS, Supelco®
Catalog # 39468-10X10MG-
Supplier:  Merck
CAS Number:  
Irritant
ɑ-Cyano-4-hydroxycinnamic acid, Ultrapure matrix substance for MALDI-MS, Supelco®
Catalog # 39468-10X10MG-
Supplier:  Merck
CAS Number:  

Specifications

  • Pk:
    10 mg
  • Seq:
    2275

Specifications

About this item

 :
 :189.17 g/mol
Melting Pt:245…250 °C (lit.)
 :GHS07
CAS Number:28166-41-8
Formula:C₁₀H₇NO₃
MDL Number:MFCD00004204