Order Entry
Austria
ContactUsLinkComponent
Acetonitrile with 0,1% ammonium acetate tested for UHPLC-MS, Supelco®
Flammable
Irritant
Acetonitrile with 0,1% ammonium acetate tested for UHPLC-MS, Supelco®
Catalog # 14274-2L
Supplier:  Merck
CAS Number:  
undefined
Flammable
Irritant
Acetonitrile with 0,1% ammonium acetate tested for UHPLC-MS, Supelco®
Catalog # 14274-2L
Supplier:  Merck
CAS Number:  

Specifications

  • Pk:
    2 L
  • Seq:
    5086

Specifications

About this item

 :
UN:1648
ADR:3,II
 :GHS02
 :GHS07
 :75-05-8