Order Entry
Austria
ContactUsLinkComponent
 
Dimethyl sulphoxide ≥99.99995% (metals basis) for inorganic trace analysis, Supelco®
Catalog # 01934-1L
Supplier:  Merck
   Safer Human and Environmental Health
CAS Number:  
undefined
Dimethyl sulphoxide ≥99.99995% (metals basis) for inorganic trace analysis, Supelco®
Catalog # 01934-1L
Supplier:  Merck
   Safer Human and Environmental Health
CAS Number:  

Specifications

  • Pk:
    1 L
  • Seq:
    5037
  • Environmentally Preferable:
    Safer Human and Environmental Health
  • Environmental certification:
    Chem21 Solvent Selection Guide recommended by default

Specifications

About this item

 :
 :78.14 g/mol
CAS Number:67-68-5
Formula:(CH₃)₂SO
MDL Number:MFCD00002089