Order Entry
Austria
ContactUsLinkComponent
FastGro synthetic, animal free, chemically defined FBS replacement, sterile, MP Biomedicals
FastGro synthetic, animal free, chemically defined FBS replacement, sterile, MP Biomedicals
Supplier:  MP Biomedicals
Certificates
undefined
FastGro synthetic, animal free, chemically defined FBS replacement, sterile, MP Biomedicals
Supplier:  MP Biomedicals

Specifications

  • pH:
    7-8 (as delivered)
  • Solubility:
    Soluble in water
  • Storage temperature:
    2…8 °C
  • Physical form:
    Light opalescent liquid

Specifications

Related Information

Frequently Bought Together