Order Entry
Austria
ContactUsLinkComponent
Isohexane (mixture of isomers) ≥95% (by anhydrous basis), AnalaR NORMAPUR® ACS analytical reagent
Isohexane (mixture of isomers) ≥95% (by anhydrous basis), AnalaR NORMAPUR® ACS analytical reagent
Supplier:  VWR Chemicals
Certificates
undefined
Isohexane (mixture of isomers) ≥95% (by anhydrous basis), AnalaR NORMAPUR® ACS analytical reagent
Supplier:  VWR Chemicals
Certificates

Specification Test Results

  • Assay (on anhydrous substance):
    min. 95.0%
  • Acidity:
    max. 0.0003 meq/g
  • Evaporation residue:
    max. 0.001%
  • Water:
    max. 0.01%
  • n-Hexane:
    max. 5%
  • Aromatics (as C6H6):
    max. 0.01%
  • Sulphur compounds (as S):
    max. 0.005%
  • Thiophene:
    max. 0.0001%
  • Substances darkened by H2SO4:
    max. 10 APHA
  • Al (Aluminium):
    max. 0.5 ppm
  • Ba (Barium):
    max. 0.1 ppm
  • Ca (Calcium):
    max. 0.5 ppm
  • Cd (Cadmium):
    max. 0.05 ppm
  • Co (Cobalt):
    max. 0.02 ppm
  • Cr (Chromium):
    max. 0.02 ppm
  • Cu (Copper):
    max. 0.02 ppm
  • Fe (Iron):
    max. 0.1 ppm
  • Mg (Magnesium):
    max. 0.1 ppm
  • Mn (Manganese):
    max. 0.02 ppm
  • Ni (Nickel):
    max. 0.02 ppm
  • Pb (Lead):
    max. 0.1 ppm
  • Sn (Tin):
    max. 0.1 ppm
  • Zn (Zinc):
    max. 0.1 ppm

Specification Test Results

Related Information

Frequently Bought Together