Order Entry
Austria
ContactUsLinkComponent
Carbon disulphide ≥99.5%, GPR RECTAPUR®
Flammable
Health
Irritant
Carbon disulphide ≥99.5%, GPR RECTAPUR®
Artikelnr. 85974.290
Supplier:  VWR Chemicals
CAS Number:  
undefined
Flammable
Health
Irritant
Carbon disulphide ≥99.5%, GPR RECTAPUR®
Artikelnr. 85974.290
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    1 L
  • Seq:
    5037

Specifications

Specification Test Results

  • Assay (G.C.) :
    min. 99,5 %
  • Identity (IR-spectrum) :
    passes test
  • Density (20°/4°) :
    1,262 - 1,264
  • Sulfates (SO4) :
    max. 0,0005 %
  • Sulfites (as SO2) :
    max. 0,003%
  • Copper (Cu) :
    max. 0,2 ppm
  • Iron (Fe) :
    max. 0,5 ppm
  • Lead (Pb) :
    max. 0,2 ppm
  • Nickel (Ni) :
    max. 0,2 ppm
  • Benzene (G.C.) :
    max. 0,005 %
  • Toluene (G.C.) :
    max. 0,005 %
  • Residue on evaporation :
    max. 0,002 %
  • Water (K.F.) :
    max. 0,02 %

Specification Test Results

About this item

 :
 :76.14 g/mol
Merck Index:13,01821
Boiling Pt:46.2 °C (1013 hPa)
Melting Pt:–111.6 °C
Density:1.2632 g/cm³ (20 °C)
Flash Pt:–30 °C
EINECS:200-843-6
UN:1131
ADR:3,I
 :GHS02
 :GHS08
 :GHS07
Storage Temperature:Ambient
CAS Number:75-15-0
Formula:CS₂
MDL Number:MFCD00011321