Sorbent tubes for gas and vapor sampling, ORBO™
Supplier: Merck
Certificates
About this item
ORBO™ sorbent tubes contain two beds of the same selective adsorbent seperated by glass wool or foam, for gas and vapor sampling.
- Wide range of high quality adsorbents
- Consistent adsorbent mesh sizes
- Coated and specially-treated materials
- Reproducible recoveries and low standard deviation
- Consistent bed weights from tube to tube to ensure capacity
The dual-layer or two-bed construction of the tube allows for any sample breakthough to be captured in the smaller back-up bed.
Frequently Bought Together