Order Entry
Austria
ContactUsLinkComponent
VWR®, Taq DNA polymerase, 200000 U
VWR®, Taq DNA polymerase, 200000 U
Catalog # 733-2479
Supplier:  VWR Chemicals
CAS Number:  
undefined
VWR®, Taq DNA polymerase, 200000 U
Catalog # 733-2479
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Concentration:
    5 U/μL
  • Pk:
    1 KIT
  • Application:
    Standard amplification
  • Format:
    Enzyme with/without buffer
  • High fidelity:
    No
  • Hot start:
    No
  • Description:
    Taq DNA Polymerase without 10X Key Buffer or 10X Extra Buffer
  • Seq:
    16
  • Storage conditions:
    – 20 °C (long term), + 4 °C (6 months)
  • Amplicon size:
    ≤ 5kb
  • Processivity:
    Standard
  • Exonuclease activity:
    5’→3’
  • Product overhang:
    3'A
  • Buffer composition:
    No Buffer
  • Concentration:
    5U/µl
  • Description:
    Taq DNA Polymerase 5U/µl 200.000 units
  • Size:
    40 x 1 ml
  • Units:
    2000000
  • Units:
    200,000 units

Specifications

About this item

Taq DNA polymerase is a thermostable recombinant DNA polymerase, which exhibits very high activity in primer extension and other molecular biology applications. The enzyme is isolated from thermus aquaticus and has a molecular weight of approximately 94 kDa.

  • Store at –20 °C

Enzyme is supplied in 20 mM tris-HCl pH 8,5, 100 mM KCl, 0,1 mM EDTA, 1 mM DTT, 0,5% tween 20, 0,5% NP40, 50% glycerol.

Caution: Reagent for in vitro laboratory use only.