Order Entry
Austria
ContactUsLinkComponent
Sodium diphenylamine-4-sulphonate, AnalaR NORMAPUR® ACS redox indicator
Sodium diphenylamine-4-sulphonate, AnalaR NORMAPUR® ACS redox indicator
Catalog # 86161.100
Supplier:  VWR Chemicals
CAS Number:  
undefined
Sodium diphenylamine-4-sulphonate, AnalaR NORMAPUR® ACS redox indicator
Catalog # 86161.100
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Pk:
    10 g
  • Seq:
    3236

Specifications

Specification Test Results

  • Sensitivity as indicator:
    Passes test
  • Loss on drying (105°C):
    ≤ 5,0

Specification Test Results

About this item

 :
 :271.27 g/mol
CAS Number:6152-67-6
Formula:C₁₂H₁₀NNaO₃S
MDL Number:MFCD00036468