VWR® TITRINORM®, pH Buffers, for Karl Fischer Titrations
85497.260
:
- Pk:1
- Description:Acid buffer, 5 mmol acid/ml
The volumetric and coulometric reagents are buffered systems, stabilised around the ideal value between 5 to 7 ensuring a rapid and stoichiometric course of reaction. Buffering systems which neutralise and stabilise the pH during the reaction are suitable for this purpose.